Learn More
Grainger Edwards Signaling Vibrating Bell, Steel, 6V DC, 4 in., 1 ea.
Catalog No. 502790604
Encompass_Preferred
This vibrating bell is designed for timing, scheduling, and general alarm applications. Engineered for indoor or outdoor use, this unit features a 102 mm (4 in.) diameter steel gong finished with corrosion-resistant heat-flowed epoxy. The striker provides continuous rapid-fire ringing as long as current is applied.
  • 6 V DC operating voltage
  • 90 dB sound rating at 3.05 m (10 ft.)
  • Surface mount design compatible with multiple electrical box sizes
  • Die-cast housing for durability
  • Self-compensating solenoid plunger
  • UL listed and FM approved
Catalog No. 50-279-0604 Supplier Grainger Supplier No. 16G574
This vibrating bell is designed for timing, scheduling, and general alarm applications. Engineered for indoor or outdoor use, this unit features a 102 mm (4 in.) diameter steel gong finished with corrosion-resistant heat-flowed epoxy. The striker provides continuous rapid-fire ringing as long as current is applied.
  • 6 V DC operating voltage
  • 90 dB sound rating at 3.05 m (10 ft.)
  • Surface mount design compatible with multiple electrical box sizes
  • Die-cast housing for durability
  • Self-compensating solenoid plunger
  • UL listed and FM approved