Learn More
Bellco Glass, Inc. Bell-Flo Complete,100mL-L32
SDP
Catalog No. 501945100
Encompass_Preferred
•Material: Type 1 Class A Borosilicate Glass. MADE IN THE USA•Dimensions: 63.5mm OD x 135mm Overall Height•Graduations: 20mL – 100mL, every 20mL•Bottom indents and three vertical baffles for optimal mixing•Two 32mm Side arms angled 45° off center for Port Assemblies•One 70mm Screw Thread Center Neck•Rugged stainless steel/ PTFE Internal Bearing Assembly•Supply with: Two 32mm PP Blue Screw Caps
Catalog No. 50-194-5100 Supplier Bellco Glass, Inc. Supplier No. 196583105
•Material: Type 1 Class A Borosilicate Glass. MADE IN THE USA•Dimensions: 63.5mm OD x 135mm Overall Height•Graduations: 20mL – 100mL, every 20mL•Bottom indents and three vertical baffles for optimal mixing•Two 32mm Side arms angled 45° off center for Port Assemblies•One 70mm Screw Thread Center Neck•Rugged stainless steel/ PTFE Internal Bearing Assembly•Supply with: Two 32mm PP Blue Screw Caps