Learn More
Bellco Glass, Inc. Bell-Flo Complete,250mL-L32
SDP
Catalog No. 501945102
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing.. MADE IN THE USA Sidearm cap: 32mm Center cap: 70mm Flask size: 250ml
Catalog No. 50-194-5102 Supplier Bellco Glass, Inc. Supplier No. 196583255
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing.. MADE IN THE USA Sidearm cap: 32mm Center cap: 70mm Flask size: 250ml