Learn More
Bellco Glass, Inc. Bell-Flo Complete, 100mL
SDP
Catalog No. 501945081
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA. Bottom indents & three vertical baffles for optimal mixing 32mm or 45mm sidearms for port assemblies Rugged stainless steel/PTFEinternal bearing assemblies. CENTER CAP:70MM, SIDEARM CAP:32MM, -OAH X OD:150 X 65
Catalog No. 50-194-5081 Supplier Bellco Glass, Inc. Supplier No. 196580105
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA. Bottom indents & three vertical baffles for optimal mixing 32mm or 45mm sidearms for port assemblies Rugged stainless steel/PTFEinternal bearing assemblies. CENTER CAP:70MM, SIDEARM CAP:32MM, -OAH X OD:150 X 65