Learn More
Bellco Glass, Inc. Bell-Flo Complete, 250mL
SDP
Catalog No. 501945086
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA. Sidearm cap: 32mm Center cap: 70mm Dimensions: 150" x 85" Flask size: 250ml
Catalog No. 50-194-5086 Supplier Bellco Glass, Inc. Supplier No. 196580255
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA. Sidearm cap: 32mm Center cap: 70mm Dimensions: 150" x 85" Flask size: 250ml