Learn More
Bellco Glass, Inc. Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. Sidearm cap: 45mm Center cap: 100mm Dimensions: 295" x 165" Flask size: 3l
SDP
Catalog No. 501945098
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. Sidearm cap: 45mm Center cap: 100mm Dimensions: 295" x 165" Flask size: 3l
Catalog No. 50-194-5098 Supplier Bellco Glass, Inc. Supplier No. 196583005
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. Sidearm cap: 45mm Center cap: 100mm Dimensions: 295" x 165" Flask size: 3l