Learn More
Bellco Glass, Inc. Bell-Flo Complete,500mL-100
SDP
Catalog No. 501945089
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA. Sidearm cap: 45mm Center cap: 100mm Dimensions: 170" x 110" Flask size: 500ml
Catalog No. 50-194-5089 Supplier Bellco Glass, Inc. Supplier No. 196580515
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA. Sidearm cap: 45mm Center cap: 100mm Dimensions: 170" x 110" Flask size: 500ml