Learn More
Bellco Glass, Inc. Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. Sidearm cap: 45mm Center cap: 70mm Dimensions: 170" x 110" Flask size: 500ml
SDP
Catalog No. 501945092
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. Sidearm cap: 45mm Center cap: 70mm Dimensions: 170" x 110" Flask size: 500ml
Catalog No. 50-194-5092 Supplier Bellco Glass, Inc. Supplier No. 196580575
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. Sidearm cap: 45mm Center cap: 70mm Dimensions: 170" x 110" Flask size: 500ml