Learn More
Bellco Glass, Inc. Bell-Flo Flask,100mL No Ctr Cap
SDP
Catalog No. 501945079
Encompass_Preferred
Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA.Specifications:Material: Type 1 Class A Borosilicate GlassSidearm Cap: 32mmCenter Cap: 70mmDimensions: 135″ X 65″Flask Size: 100ml
Catalog No. 50-194-5079 Supplier Bellco Glass, Inc. Supplier No. 196580100
Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA.Specifications:Material: Type 1 Class A Borosilicate GlassSidearm Cap: 32mmCenter Cap: 70mmDimensions: 135″ X 65″Flask Size: 100ml