Learn More
Bellco Glass, Inc. Bell-Flo Flask, 1L No Ctr Cap
SDP
Catalog No. 501945093
Encompass_Preferred
1000mL Bell-Flo Spinner Flask. MADE IN THE USAFlask Specifications:Material: Type 1 Class A Borosilicate GlassDimensions: 135mm OD x 240mm Overall Height, 250mm across sidearmsTwo 45mm sidearms angled 60⁰ off the centerlineOne 100mm Screw Thread Center NeckIndented Bottom: Prevents dead spot under impellerSide Baffles: Three equally spaced baffles, 80mm long x 9mm deepSupply with: Two 45mm PP Blue Screw Caps & PP Blue Pouring Rings
Catalog No. 50-194-5093 Supplier Bellco Glass, Inc. Supplier No. 196581000
1000mL Bell-Flo Spinner Flask. MADE IN THE USAFlask Specifications:Material: Type 1 Class A Borosilicate GlassDimensions: 135mm OD x 240mm Overall Height, 250mm across sidearmsTwo 45mm sidearms angled 60⁰ off the centerlineOne 100mm Screw Thread Center NeckIndented Bottom: Prevents dead spot under impellerSide Baffles: Three equally spaced baffles, 80mm long x 9mm deepSupply with: Two 45mm PP Blue Screw Caps & PP Blue Pouring Rings