Learn More
Bellco Glass, Inc. Bell-Flo Flask, 250mL No Ctr Cap
SDP
Catalog No. 501945084
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA.Sidearm cap: 32mmCenter cap: 70mmDimensions: 140″ x 85″Flask size: 250ml
Catalog No. 50-194-5084 Supplier Bellco Glass, Inc. Supplier No. 196580250
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA.Sidearm cap: 32mmCenter cap: 70mmDimensions: 140″ x 85″Flask size: 250ml