Learn More
Bellco Glass, Inc. Bell-Flo Flask,250mL-L32 No Ctr Cap
SDP
Catalog No. 501945101
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA. Sidearm cap: 32mm Center cap: 70mm Flask size: 250ml
Catalog No. 50-194-5101 Supplier Bellco Glass, Inc. Supplier No. 196583250
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA. Sidearm cap: 32mm Center cap: 70mm Flask size: 250ml