Learn More
Bellco Glass, Inc. Bell-Flo Flask,500mL-100 NK No Ctr Cap
SDP
Catalog No. 501945087
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA. Sidearm cap: 45mm Center cap: 100mm, sold separately Dimensions: 170" x 110" Flask size: 500ml
Catalog No. 50-194-5087 Supplier Bellco Glass, Inc. Supplier No. 196580510
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA. Sidearm cap: 45mm Center cap: 100mm, sold separately Dimensions: 170" x 110" Flask size: 500ml