Learn More
Bellco Glass, Inc. Bell-Flo Flask, 500mL-70mm No Ctr Cap
SDP
Catalog No. 501945090
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA. Sidearm cap: 45mm Center cap: 70mm sold separately Dimensions: 1110" x 175" Flask size: 500ml
Catalog No. 50-194-5090 Supplier Bellco Glass, Inc. Supplier No. 196580570
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. MADE IN THE USA. Sidearm cap: 45mm Center cap: 70mm sold separately Dimensions: 1110" x 175" Flask size: 500ml