Learn More
Bellco Glass, Inc. Bell-Flo IOB Assembly ,250mL
SDP
Catalog No. 501945085
Encompass_Preferred
Bell-Flo IOB assembly was designed to be used with the bell-flo vessel with bottom indents and 3 vertical baffles for optimal mixing. Rugged stainless steel ptfe internal bearing impeller assemblies utilize a wing style impeller blade that increases mixing. MADE IN THE USA. CENTER CAP:70MM, SIDEARM CAP:32MM, -OAH X OD:130 X 65
Catalog No. 50-194-5085 Supplier Bellco Glass, Inc. Supplier No. 196580251
Bell-Flo IOB assembly was designed to be used with the bell-flo vessel with bottom indents and 3 vertical baffles for optimal mixing. Rugged stainless steel ptfe internal bearing impeller assemblies utilize a wing style impeller blade that increases mixing. MADE IN THE USA. CENTER CAP:70MM, SIDEARM CAP:32MM, -OAH X OD:130 X 65