Learn More
Bellco Glass, Inc. Bell-Flo IOB Assembly , 3L
SDP
Catalog No. 501945097
Encompass_Preferred
3000mL Bell-Flo Internal Overhead Bearing Assembly. MADE IN THE USAInternal Overhead Bearing Assembly Specifications:Consists of screw cap, cap seal, sealing ring, compression washer, ball bearing spacer, ball bearing, bearing thimble, magnet, o-ring, magnet holder, shaft, & paddle100mm PP Blue Screw Cap: 82.6mm OD x 22.4mm tallPTFE Cap Seal: 95.3mm OD x 25mm longRed Silicone Sealing Ring: 86mm OD x 76.2mm ID x 6.1mm thickWhite Silicone Compression Washer: 27.0mm OD x 12.7mm ID x 1.6mm thickPTFE Ball Bearing Spacer: 19.1mm OD x 12.7mm ID x 22.4mm deepPBT & SST Ball Bearing: 19.1mm OD x 6.4mm ID x 7.1mm deepPTFE Bearing Thimble: 31.8mm OD x 19.1mm ID x 38.1mm longPTFE Magnet: 12.7mm OD x 76.2mm longSilicone O-ring: 7.9mm OD x 4.8mm IDPTFE Magnet Holder: 19.1mm OD x 177.8mm long316L SST Shaft: 6.4mm OD x 190.5mm longPTFE Paddle: 82.6mm wide x 177.8mm long x 1.6mm thick
Catalog No. 50-194-5097 Supplier Bellco Glass, Inc. Supplier No. 196583001
3000mL Bell-Flo Internal Overhead Bearing Assembly. MADE IN THE USAInternal Overhead Bearing Assembly Specifications:Consists of screw cap, cap seal, sealing ring, compression washer, ball bearing spacer, ball bearing, bearing thimble, magnet, o-ring, magnet holder, shaft, & paddle100mm PP Blue Screw Cap: 82.6mm OD x 22.4mm tallPTFE Cap Seal: 95.3mm OD x 25mm longRed Silicone Sealing Ring: 86mm OD x 76.2mm ID x 6.1mm thickWhite Silicone Compression Washer: 27.0mm OD x 12.7mm ID x 1.6mm thickPTFE Ball Bearing Spacer: 19.1mm OD x 12.7mm ID x 22.4mm deepPBT & SST Ball Bearing: 19.1mm OD x 6.4mm ID x 7.1mm deepPTFE Bearing Thimble: 31.8mm OD x 19.1mm ID x 38.1mm longPTFE Magnet: 12.7mm OD x 76.2mm longSilicone O-ring: 7.9mm OD x 4.8mm IDPTFE Magnet Holder: 19.1mm OD x 177.8mm long316L SST Shaft: 6.4mm OD x 190.5mm longPTFE Paddle: 82.6mm wide x 177.8mm long x 1.6mm thick