Learn More
Bellco Glass, Inc. Bell-Stirr 4 Position, 100V -500mL
SDP
Catalog No. 501946190
Encompass_Preferred
Bellco Magnetic Stirrers are specifically designed for growing both mammalian and bacterial cultures where minimal heat transfer from the magnetic stirrer to the culture vessel is essential. They are ideal for tissue cultures grown in spinner flasks. Bellco incorporates low wattage DC motors to reduce stirrer heat up. Independent power and speed controls allow unit to maintain a speed setting when switched on and off. Powerful magnets rotate on ball bearing shafts for increased coupling and continuous maintenance free service. Polyurethane drive belts resist humidity and temperature. The 100V, 115V & 230V models feature a convenience outlet that allows daisy chaining of up to ten stirrers. . MADE IN THE USASpeed range: 30 - 1000 rpm +/- 5%
Catalog No. 50-194-6190 Supplier Bellco Glass, Inc. Supplier No. 776006105
Bellco Magnetic Stirrers are specifically designed for growing both mammalian and bacterial cultures where minimal heat transfer from the magnetic stirrer to the culture vessel is essential. They are ideal for tissue cultures grown in spinner flasks. Bellco incorporates low wattage DC motors to reduce stirrer heat up. Independent power and speed controls allow unit to maintain a speed setting when switched on and off. Powerful magnets rotate on ball bearing shafts for increased coupling and continuous maintenance free service. Polyurethane drive belts resist humidity and temperature. The 100V, 115V & 230V models feature a convenience outlet that allows daisy chaining of up to ten stirrers. . MADE IN THE USASpeed range: 30 - 1000 rpm +/- 5%