Learn More
Electron Microscopy Sciences Bellco Brand Cover Slips, 9x9mm, 100/Pack
SDP
Catalog No. 50949006
Encompass_Preferred
For use with Bellco Leighton Tissue Culture Tubes. Made of clear white borosilicate glass. #1 thickness (0.13 to 0.17 mm). Packed one ounce (28 g) per case.Size: 9 x 9 mmPackaging: 100 pieces/oz
Catalog No. 50-949-006 Supplier Electron Microscopy Sciences Supplier No. 7219009
For use with Bellco Leighton Tissue Culture Tubes. Made of clear white borosilicate glass. #1 thickness (0.13 to 0.17 mm). Packed one ounce (28 g) per case.Size: 9 x 9 mmPackaging: 100 pieces/oz