Learn More
Bellco Glass, Inc. Bellco Heating Blanket,115V-36L 1200 Watts
SDP
Catalog No. 501944814
Encompass_Preferred
Heating blankets are designed to be used with a temperature controller and rtd temperature sensor for heating media inside a vessel to temperature. MADE IN THE USA. Male IEC connector for use with temperature controllersHeating at 5 watts/sq.in.Flask size: 36lWatts: 1200 **not for celltrol** use 600 watt blanket (1964-57365)Dimension (in.): 5 x 48 3/4"3 latches6ft cord (18-2) with cj14 plugCaution: vessel must be stirred when heating blanket is in operation
Catalog No. 50-194-4814 Supplier Bellco Glass, Inc. Supplier No. 196455360
Heating blankets are designed to be used with a temperature controller and rtd temperature sensor for heating media inside a vessel to temperature. MADE IN THE USA. Male IEC connector for use with temperature controllersHeating at 5 watts/sq.in.Flask size: 36lWatts: 1200 **not for celltrol** use 600 watt blanket (1964-57365)Dimension (in.): 5 x 48 3/4"3 latches6ft cord (18-2) with cj14 plugCaution: vessel must be stirred when heating blanket is in operation