Learn More
Bellco Glass, Inc. Bellco Heating Blanket,115V-3L-Bio 130 Watts-Bioreator Vessel
SDP
Catalog No. 501944802
Encompass_Preferred
Heating Blankets are designed to be used with a temperature controller and RTD temperature sensor for heating media inside a vessel to temperature. MADE IN THE USA. Specifications:Flask Size: 3l Bioreactor Vessel (1964-88403)Dimensions: 15″ X 2.5″ Tall With 1 LatchWatts: 130 Heating @ 5 Watts Per Sq In Male IEC Connector For Use With Temperature Controller6ft Cord (18-2) With CJ14 Plug
Catalog No. 50-194-4802 Supplier Bellco Glass, Inc. Supplier No. 196451003
Heating Blankets are designed to be used with a temperature controller and RTD temperature sensor for heating media inside a vessel to temperature. MADE IN THE USA. Specifications:Flask Size: 3l Bioreactor Vessel (1964-88403)Dimensions: 15″ X 2.5″ Tall With 1 LatchWatts: 130 Heating @ 5 Watts Per Sq In Male IEC Connector For Use With Temperature Controller6ft Cord (18-2) With CJ14 Plug