Learn More
Bellco Glass, Inc. Bellco Heating Blanket,115V-3L-LW
SDP
Catalog No. 501944829
Encompass_Preferred
Heating blankets are designed to be used with a temperature controller and rtd temperature sensor for heating media inside a vessel to temperature. MADE IN THE USA. Heating blanket - benchmark Flask size: 3lWatts: 295Heating @ 5 watts per sq inMale IEC connector for use with temperature controllerDimensions: 2" x 29.5"1 latch6 ft cord (18-2) with cj14 plugCaution: vessel must be stirred when heating blanket is in operation
Catalog No. 50-194-4829 Supplier Bellco Glass, Inc. Supplier No. 196459003
Heating blankets are designed to be used with a temperature controller and rtd temperature sensor for heating media inside a vessel to temperature. MADE IN THE USA. Heating blanket - benchmark Flask size: 3lWatts: 295Heating @ 5 watts per sq inMale IEC connector for use with temperature controllerDimensions: 2" x 29.5"1 latch6 ft cord (18-2) with cj14 plugCaution: vessel must be stirred when heating blanket is in operation