Learn More
Bellco Glass, Inc. Bellco Heating Blanket,230V-1L 75 Watts
SDP
Catalog No. 501944816
Encompass_Preferred
Heating blankets are designed to be used with a temperature controller and rtd temperature sensor for heating media inside a vessel to temperature. MADE IN THE USA. Male IEC connector for use with temperature controllersHeating at 5 watts/sq.in.Flask size: 1lWatts: 75Dimension (in.): 1 x 13 3/4"1 latch6ft cord (18-2) with cj14 plugCaution: vessel must be stirred when heating blanket is in operation
Catalog No. 50-194-4816 Supplier Bellco Glass, Inc. Supplier No. 196456010
Heating blankets are designed to be used with a temperature controller and rtd temperature sensor for heating media inside a vessel to temperature. MADE IN THE USA. Male IEC connector for use with temperature controllersHeating at 5 watts/sq.in.Flask size: 1lWatts: 75Dimension (in.): 1 x 13 3/4"1 latch6ft cord (18-2) with cj14 plugCaution: vessel must be stirred when heating blanket is in operation