Learn More
Bellco Glass, Inc. Bellco Heating Blanket,230V-36L-LW 375 Watts - Low Wattage
SDP
Catalog No. 501944827
Encompass_Preferred
Heating blankets are designed to be used with a temperature controller and rtd temperature sensor for heating media inside a vessel to temperature. MADE IN THE USA. Male IEC connector for use with temperature controllersHeating at 5 watts/sq.in.Flask size: 36lWatts: 375 - low wattageDimension (in.): 5 x 47 3/43 latches6ft cord (18-2) with cj14 plugCaution: vessel must be stirred when heating blanket is in operation
Catalog No. 50-194-4827 Supplier Bellco Glass, Inc. Supplier No. 196458360
Heating blankets are designed to be used with a temperature controller and rtd temperature sensor for heating media inside a vessel to temperature. MADE IN THE USA. Male IEC connector for use with temperature controllersHeating at 5 watts/sq.in.Flask size: 36lWatts: 375 - low wattageDimension (in.): 5 x 47 3/43 latches6ft cord (18-2) with cj14 plugCaution: vessel must be stirred when heating blanket is in operation