Learn More
Bellco Glass, Inc. Bellco Heating Blanket,230V-8L 375 Watts
SDP
Catalog No. 501944819
Encompass_Preferred
Heating blankets are designed to be used with a temperature controller and rtd temperature sensor for heating media inside a vessel to temperature. MADE IN THE USA. Male IEC connector for use with temperature controllersHeating at 5 watts/sq.in.Flask size: 8lWatts: 375Dimension (in.): 3 x 26.52 latches6ft cord (18-2) with cj14 plugCaution: vessel must be stirred when heating blanket is in operation
Catalog No. 50-194-4819 Supplier Bellco Glass, Inc. Supplier No. 196456080
Heating blankets are designed to be used with a temperature controller and rtd temperature sensor for heating media inside a vessel to temperature. MADE IN THE USA. Male IEC connector for use with temperature controllersHeating at 5 watts/sq.in.Flask size: 8lWatts: 375Dimension (in.): 3 x 26.52 latches6ft cord (18-2) with cj14 plugCaution: vessel must be stirred when heating blanket is in operation