Learn More
Bellco Glass, Inc. The motion alarm produces an audible alarm should the drive mechanism fail in anyway. The alarm reset automatically upon resumption of motion and may be silenced with an override button on the back panel. The alarms are factory installed only in our proce
SDP
Catalog No. 501946223
Encompass_Preferred
The motion alarm produces an audible alarm should the drive mechanism fail in anyway. The alarm reset automatically upon resumption of motion and may be silenced with an override button on the back panel. The alarms are factory installed only in our proce
Catalog No. 50-194-6223 Supplier Bellco Glass, Inc. Supplier No. 778522222
The motion alarm produces an audible alarm should the drive mechanism fail in anyway. The alarm reset automatically upon resumption of motion and may be silenced with an override button on the back panel. The alarms are factory installed only in our proce