Learn More
Waters Corp Waters Belleville Spring Washer, .25 in x .125 in, 10/pk
Catalog No. 50792830 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Belleville spring washer is a crucial replacement component for a variety of analytical instruments. Measuring .25 in x .125 in, it is supplied in packs of 10 units. This washer is integral for maintaining secure seals in systems, particularly in applications such as Xevo mass spectrometers, where it helps prevent potential source leaks.
  • Replacement component for analytical instruments.
  • Dimensions: .25 in x .125 in.
  • Supplied in packs of 10 units.
  • Helps prevent source leaks in mass spectrometry systems.
  • Compatible with column heaters, ELS detectors, and other detector types.
  • Suitable for HPLC, UPLC, and MS system applications.
  • Supports a broad range of scientific instrumentation.
Catalog No. 50-792-830 Supplier Waters Corp Supplier No. 700003080
This Belleville spring washer is a crucial replacement component for a variety of analytical instruments. Measuring .25 in x .125 in, it is supplied in packs of 10 units. This washer is integral for maintaining secure seals in systems, particularly in applications such as Xevo mass spectrometers, where it helps prevent potential source leaks.
  • Replacement component for analytical instruments.
  • Dimensions: .25 in x .125 in.
  • Supplied in packs of 10 units.
  • Helps prevent source leaks in mass spectrometry systems.
  • Compatible with column heaters, ELS detectors, and other detector types.
  • Suitable for HPLC, UPLC, and MS system applications.
  • Supports a broad range of scientific instrumentation.