Learn More
Electron Microscopy Sciences Bellows Dropping Bottle 250 ml, 60 mm diameter, 136 mm height
SDP
Catalog No. 5024659
Encompass_Preferred
A low density polyethylene (LDPE) bottle supplied with a removable bellow-type dropping pipette.  The pipettes are graduated for nominal indication of amount of liquid being drawn.  Bellows-shaped bulb makes for easy use and provides excellent drop information.
Diameter: 60 mm
Height: 136 mm
Cap: 24 mm
Catalog No. 50-246-59 Supplier Electron Microscopy Sciences Supplier No. 6429808
A low density polyethylene (LDPE) bottle supplied with a removable bellow-type dropping pipette.  The pipettes are graduated for nominal indication of amount of liquid being drawn.  Bellows-shaped bulb makes for easy use and provides excellent drop information.
Diameter: 60 mm
Height: 136 mm
Cap: 24 mm