Learn More
Electron Microscopy Sciences Bellows Dropping Bottle 60 ml, 38 mm diameter, 86 mm height
SDP
Catalog No. 5024657
Encompass_Preferred
A low density polyethylene (LDPE) bottle supplied with a removable bellow-type dropping pipette.  The pipettes are graduated for nominal indication of amount of liquid being drawn.  Bellows-shaped bulb makes for easy use and provides excellent drop information.
Diameter: 38 mm
Height: 86 mm
Cap: 18 mm
Catalog No. 50-246-57 Supplier Electron Microscopy Sciences Supplier No. 6429802
A low density polyethylene (LDPE) bottle supplied with a removable bellow-type dropping pipette.  The pipettes are graduated for nominal indication of amount of liquid being drawn.  Bellows-shaped bulb makes for easy use and provides excellent drop information.
Diameter: 38 mm
Height: 86 mm
Cap: 18 mm