Learn More
Sigma Aldrich Fine Chemicals Biosciences Belly Dancer/Hybridization1EA
Catalog No. 502908872
Encompass
The Belly Dancer/Hybridization water bath offers ideal benchtop temperature control and orbital mixing in one versatile unit. The lightweight thermostat-controlled water bath is designed for use on the Belly Dancer platform and its controls are integrated into the Belly Dancer. The orbital hybridization water bath is the ideal instrument for Northern and Southern blots high stringency filter washing in situ hybridization and hot staining and destaining protocols.
Catalog No. 50-290-8872 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3676211EA
The Belly Dancer/Hybridization water bath offers ideal benchtop temperature control and orbital mixing in one versatile unit. The lightweight thermostat-controlled water bath is designed for use on the Belly Dancer platform and its controls are integrated into the Belly Dancer. The orbital hybridization water bath is the ideal instrument for Northern and Southern blots high stringency filter washing in situ hybridization and hot staining and destaining protocols.