Learn More
Medchemexpress LLC HY-13205 50mg , Belnacasan CAS:273404-37-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1320550MG
Encompass_Preferred
HY-13205 50mg Belnacasan CAS: 273404-37-8 Belnacasan (VX-765) is an orally bioactive prodrug of VRT-043198, which is a potent and selective inhibitor of IL-converting enzyme (ICE)/caspase-1 with Kis of 0.8 nM and less than 0.6 nM for caspase-1 and caspase-4, respectively. Belnacasan (VX-765) inhibits the release of LPS-induced IL-1β and IL-18 by human PBMCs with an IC50 of ~0.7 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0041