Learn More
Selleck Chemical LLC Belnacasan (VX-765) S2228-50mg
SDP
Catalog No. 503108177
Encompass_Preferred
Belnacasan (VX-765) is a potent and selective inhibitor of caspase-1 with Ki of 0 8 nM in a cell-free assay Phase 2
Catalog No. 50-310-8177 Supplier Selleck Chemical LLC Supplier No. S222850MG
Add to Cart
Add to Cart
Belnacasan (VX-765) is a potent and selective inhibitor of caspase-1 with Ki of 0 8 nM in a cell-free assay Phase 2