Learn More
Selleck Chemical LLC Belnacasan (VX-765) S2228-200mg
SDP
Catalog No. 503112674
Encompass_Preferred
Belnacasan (VX-765) is a potent and selective inhibitor of caspase-1 with Ki of 0 8 nM in a cell-free assay Phase 2
Catalog No. 50-311-2674 Supplier Selleck Chemical LLC Supplier No. S2228200MG
Belnacasan (VX-765) is a potent and selective inhibitor of caspase-1 with Ki of 0 8 nM in a cell-free assay Phase 2