Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Belotecan-d7 hydrochloride | 1346598-22-8 | 99.9% | 477.00 | C25H21D7ClN3O4 | 5 MG
SDP

Catalog No. 5000222889
Encompass_Preferred

Belotecan-d7 hydrochloride is a deuterium-labeled analog of belotecan hydrochloride provided as a stable-isotope reference standard for analytical and research applications. It is intended for use as an internal standard in mass spectrometry, ADME, and pharmacokinetic studies to support accurate quantitation and compound identification.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (99.9%) suitable for analytical workflows.
  • Supplied as the hydrochloride salt for improved stability and solubility.
  • Molecular weight 477.00 and formula C25H21D7ClN3O4 for precise mass tagging.
  • Available in small pack sizes appropriate for analytical use.

Catalog No. 50-002-22889 Supplier Medchemexpress LLC Supplier No. HY13566AS5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Belotecan-d7 hydrochloride is a deuterium-labeled analog of belotecan hydrochloride provided as a stable-isotope reference standard for analytical and research applications. It is intended for use as an internal standard in mass spectrometry, ADME, and pharmacokinetic studies to support accurate quantitation and compound identification.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (99.9%) suitable for analytical workflows.
  • Supplied as the hydrochloride salt for improved stability and solubility.
  • Molecular weight 477.00 and formula C25H21D7ClN3O4 for precise mass tagging.
  • Available in small pack sizes appropriate for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.