Learn More
Medchemexpress LLC Belotecan hydrochloride | 213819-48-8 | MFCD07772313 | 99.0% | C25H28ClN3O4 | 10MG
SDP
Catalog No. 5000260039
Encompass_Preferred
Belotecan hydrochloride is a synthetic camptothecin derivative that acts as a potent topoisomerase I inhibitor used for preclinical oncology research. It is supplied as a solid powder and as a DMSO solution for laboratory studies, and is used to investigate DNA damage, cell cycle effects, and anticancer activity in cell-based and biochemical assays.
  • Potent topoisomerase I inhibition for anticancer research.
  • Available as solid powder and as a 10 mM solution in DMSO.
  • High reported purity suitable for preclinical assays.
  • White to yellow solid appearance facilitating visual inspection.
  • Supplied with documentation such as COA and SDS for quality and safety.
Catalog No. 50-002-60039 Supplier Medchemexpress LLC Supplier No. HY13566A10MG
Belotecan hydrochloride is a synthetic camptothecin derivative that acts as a potent topoisomerase I inhibitor used for preclinical oncology research. It is supplied as a solid powder and as a DMSO solution for laboratory studies, and is used to investigate DNA damage, cell cycle effects, and anticancer activity in cell-based and biochemical assays.
  • Potent topoisomerase I inhibition for anticancer research.
  • Available as solid powder and as a 10 mM solution in DMSO.
  • High reported purity suitable for preclinical assays.
  • White to yellow solid appearance facilitating visual inspection.
  • Supplied with documentation such as COA and SDS for quality and safety.