Learn More
Medchemexpress LLC HY-13566A 5mg Medchemexpress, Belotecan (hydrochloride) CAS:213819-48-8 Purity:>98%
SDP
Catalog No. 502081395
Encompass_Preferred
Medchemexpress, HY-13566A 5mg Belotecan (hydrochloride) CAS:213819-48-8 Belotecan hydrochloride (CKD-602 hydrochloride), a Topoisomerase I inhibitor, is a synthetic and water-soluble camptothecin derivative. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1395 Supplier Medchemexpress LLC Supplier No. HY13566A5MG
Medchemexpress, HY-13566A 5mg Belotecan (hydrochloride) CAS:213819-48-8 Belotecan hydrochloride (CKD-602 hydrochloride), a Topoisomerase I inhibitor, is a synthetic and water-soluble camptothecin derivative. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.