Learn More
Cayman Chemical BelotecnhydrochlorIde 50mg
SDP
Catalog No. 502910102
Encompass_Preferred
A DNA topoisomerase I inhibitor (IC50 = 0.119 UG/ml); a derivative of camptothecin; inhibits the proliferation of KATO III, HT-29, A549, MDA-MB-231, and SKOV3 cells (IC50s = 160, 10.9, 9, 345, and 31 ng/ml, respectively); induces apoptosis and cell cycle arrest at the G2/M phase in, and inhibits invasion of, SiHa cells at 150 ng/ml; reduces tumor growth in a Ca Ski cervical cancer mouse xenograft model at 25 mg/kg; reduces food intake and body weight in pregnant dams and increases fetal deaths and decreases litter size at 80 UG/kg
Catalog No. 50-291-0102 Supplier Cayman Chemical Supplier No. 3548850 MG
A DNA topoisomerase I inhibitor (IC50 = 0.119 UG/ml); a derivative of camptothecin; inhibits the proliferation of KATO III, HT-29, A549, MDA-MB-231, and SKOV3 cells (IC50s = 160, 10.9, 9, 345, and 31 ng/ml, respectively); induces apoptosis and cell cycle arrest at the G2/M phase in, and inhibits invasion of, SiHa cells at 150 ng/ml; reduces tumor growth in a Ca Ski cervical cancer mouse xenograft model at 25 mg/kg; reduces food intake and body weight in pregnant dams and increases fetal deaths and decreases litter size at 80 UG/kg