Learn More
Medchemexpress LLC HY-15307 5mg , Belumosudil CAS:911417-87-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY153075MG
Encompass_Preferred
HY-15307 5mg Belumosudil CAS: 911417-87-3 Belumosudil (KD025) is a selective inhibitor of ROCK2 with IC50s of 105 nM and 24 µM for ROCK2 and ROCK1, respectively. Anti-fibrotic properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9532