Learn More
Medchemexpress LLC HY-109080 5mg Medchemexpress, Belvarafenib CAS:1446113-23-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1090805MG
Encompass_Preferred
Medchemexpress, HY-109080 5mg Belvarafenib CAS:1446113-23-0 Belvarafenib (HM95573) is a potent and pan RAF (Rapidly Accelerated Fibrosarcoma) inhibitor, with IC50s of 56 nM, 7 nM and 5 nM for B-RAF, B-RAFv600E and C-RAF respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9663