Learn More
Medchemexpress LLC HY-B1541 10mg Medchemexpress, Bemesetron CAS:40796-97-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB154110MG
Encompass_Preferred
Medchemexpress, HY-B1541 10mg Bemesetron CAS:40796-97-2 Bemesetron (MDL 72222) is a selective 5-HT 3 receptor antagonist with an IC 50 of 0.33 nM [1] . Neuroprotective effect [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2966