Learn More
Medchemexpress LLC Bemnifosbuvir | 1998705-64-8 | 99.0% | 581.53 | 50 MG
SDP
Catalog No. 5000350564
Encompass_Preferred
Bemnifosbuvir (AT-511) is a potent and orally active inhibitor of HCV viral replication. It is highly effective in controlling SARS-CoV-2 (COVID-19) infection in vitro with an EC90 of 0.47 μM. Bemnifosbuvir exhibits pangenotypic antiviral activity.
  • Highly effective in controlling SARS-CoV-2 (COVID-19) infection in vitro
  • Exhibits pangenotypic antiviral activity
  • Inhibits HCV genotype 1a (EC50: 12.8 nM)
  • Inhibits HCV GT1b (EC50: 12.5 nM)
  • Inhibits HCV GT2a (EC50: 9.2 nM)
  • Inhibits HCV GT3a (EC50: 10.3 nM)
  • Inhibits HCV GT4a (EC50: 14.7 nM)
  • Inhibits HCV GT5a (EC50: 28.5 nM)
  • Preferentially delivers high levels of AT-9010 to the liver in vivo
Catalog No. 50-003-50564 Supplier Medchemexpress LLC Supplier No. HY13795850MG
Bemnifosbuvir (AT-511) is a potent and orally active inhibitor of HCV viral replication. It is highly effective in controlling SARS-CoV-2 (COVID-19) infection in vitro with an EC90 of 0.47 μM. Bemnifosbuvir exhibits pangenotypic antiviral activity.
  • Highly effective in controlling SARS-CoV-2 (COVID-19) infection in vitro
  • Exhibits pangenotypic antiviral activity
  • Inhibits HCV genotype 1a (EC50: 12.8 nM)
  • Inhibits HCV GT1b (EC50: 12.5 nM)
  • Inhibits HCV GT2a (EC50: 9.2 nM)
  • Inhibits HCV GT3a (EC50: 10.3 nM)
  • Inhibits HCV GT4a (EC50: 14.7 nM)
  • Inhibits HCV GT5a (EC50: 28.5 nM)
  • Preferentially delivers high levels of AT-9010 to the liver in vivo