Learn More
Medchemexpress LLC Bemnifosbuvir hemisulfate | 2241337-84-6 | 99.4% | C24H33FN7O7P.1/2H2O4S | 10MG
SDP
Catalog No. 5000258478
Encompass_Preferred
Bemnifosbuvir hemisulfate is the hemisulfate salt of a guanosine nucleotide prodrug developed as an orally active antiviral. It demonstrates potent activity against hepatitis C virus and in vitro activity against SARS-CoV-2 (reported EC90 ≈ 0.47 μM). Supplied for research use, the compound is provided at high purity and in laboratory-scale quantities suitable for pharmacology and mechanism-of-action studies.
  • Hemisulfate salt form for improved handling and solubility
  • High purity (≈99.4%) suitable for in vitro research
  • Orally bioactive nucleotide prodrug mechanism for antiviral studies
  • Demonstrated activity against HCV and in vitro SARS-CoV-2 models
  • Suitable for pharmacology and mechanism-of-action experiments
  • Supplied in laboratory-scale sizes for research flexibility
Catalog No. 50-002-58478 Supplier Medchemexpress LLC Supplier No. HY13795810MG
Bemnifosbuvir hemisulfate is the hemisulfate salt of a guanosine nucleotide prodrug developed as an orally active antiviral. It demonstrates potent activity against hepatitis C virus and in vitro activity against SARS-CoV-2 (reported EC90 ≈ 0.47 μM). Supplied for research use, the compound is provided at high purity and in laboratory-scale quantities suitable for pharmacology and mechanism-of-action studies.
  • Hemisulfate salt form for improved handling and solubility
  • High purity (≈99.4%) suitable for in vitro research
  • Orally bioactive nucleotide prodrug mechanism for antiviral studies
  • Demonstrated activity against HCV and in vitro SARS-CoV-2 models
  • Suitable for pharmacology and mechanism-of-action experiments
  • Supplied in laboratory-scale sizes for research flexibility