Learn More
Selleck Chemical LLC Bemnifosbuvir Hemisulfate-5MG
SDP
Catalog No. 503134524
Encompass_Preferred
Bemnifosbuvir Hemisulfate (AT-527 RG-6422 RO 7496998) is a hemi-sulfate salt of AT-511 AT-511 is a potent inhibitor of SARS-CoV-2 with an EC90 of 0 47 M
Catalog No. 50-313-4524 Supplier Selleck Chemical LLC Supplier No. E10135MG
Bemnifosbuvir Hemisulfate (AT-527 RG-6422 RO 7496998) is a hemi-sulfate salt of AT-511 AT-511 is a potent inhibitor of SARS-CoV-2 with an EC90 of 0 47 M