Learn More
Medchemexpress LLC Bemotrizinol 10g
SDP
Catalog No. 5000204859
Encompass_Preferred
Bemotrizinol (Tinosorb S) a compound that can absorb ultraviolet light has been shown to accept atomic oxygen generated by N-oxide photodeoxygenation[1][2][3][4]
Catalog No. 50-002-04859 Supplier Medchemexpress LLC Supplier No. HYW09973010G
Add to Cart
Add to Cart
Bemotrizinol (Tinosorb S) a compound that can absorb ultraviolet light has been shown to accept atomic oxygen generated by N-oxide photodeoxygenation[1][2][3][4]