Learn More
Honeywell™ Ben 2 Plus Structural Helmets
Description
Strong, high heat-resistant shell with adjustable suspension.
- FYR-glass shell
- Clear 4 × 0.150 in. Hi-Temp APEC faceshield
- Nomex™ quick-release chin strap with postman slide
- Eight Reflexite™ lime trapezoids
- Black Nomex/flannel ear covers
- Kevlar™ felt winter liner dome
HDO Model
- Black Nomex/flannel ear covers
- Black Nomex/Crosstech™ ear covers
- Eagle front holder—golden alloy plating aluminum with hardware
Order Info
Available through Fisher Safety's GSA Advantage!™ Schedule 84.
Certifications
NFPA 1971, 2007 Edition.
Warranty and Services
Lifetime warranty
Specifications
Specifications
| Certifications/Compliance | NFPA 1971, 2007 Edition. |
| Color | Black |
| Liner Material | Dupont™ Kevlar™ aramid fiber Felt Winter |
| Material | Shell: FYR-Glass; Ear Covers: Yellow Nomex FR Cotton |