Learn More
Honeywell™ Ben 2 Plus Structural Helmets
Catalog No. 19167092
Change view
Click to view available options
Color:
Black
Red
White
Yellow
Material:
Shell: FYR-Glass; Ear Covers: Yellow Nomex FR Cotton
Shell: FYR-Glass; Ear Covers: Yellow Nomex Moisture Barrier FR Cotton
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material
19-167-092 White Shell: FYR-Glass; Ear Covers: Yellow Nomex FR Cotton
19-167-140 Black Shell: FYR-Glass; Ear Covers: Yellow Nomex Moisture Barrier FR Cotton
19-167-090 Black Shell: FYR-Glass; Ear Covers: Yellow Nomex FR Cotton
19-167-143 Yellow Shell: FYR-Glass; Ear Covers: Yellow Nomex Moisture Barrier FR Cotton
19-167-141 Red Shell: FYR-Glass; Ear Covers: Yellow Nomex Moisture Barrier FR Cotton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-167-092 Supplier Honeywell™ Supplier No. SAEGSA00513
Add to Cart
Add to Cart
Rugged and dependable helmet for structural firefighting operations
Strong, high heat-resistant shell with adjustable suspension.
  • FYR-glass shell
  • Clear 4 × 0.150 in. Hi-Temp APEC faceshield
  • Nomex™ quick-release chin strap with postman slide
  • Eight Reflexite™ lime trapezoids
  • Black Nomex/flannel ear covers
  • Kevlar™ felt winter liner dome
HDO Model
  • Black Nomex/flannel ear covers
  • BPR Model
    • Black Nomex/Crosstech™ ear covers
    • Eagle front holder—golden alloy plating aluminum with hardware
    Order Info
    Available through Fisher Safety's GSA Advantage!™ Schedule 84.
    Certifications
    NFPA 1971, 2007 Edition.
    Warranty and Services
    Lifetime warranty
    TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance NFPA 1971, 2007 Edition.
    Color White
    Liner Material Dupont™ Kevlar™ aramid fiber Felt Winter
    Material Shell: FYR-Glass; Ear Covers: Yellow Nomex FR Cotton