Learn More
Selleck Chemical LLC Benactyzine hydrochloride S3701-25mg
SDP
Catalog No. 503108784
Encompass_Preferred
Benactyzine hydrochloride is a centrally acting muscarinic antagonist It is used as an antidepressant in the treatment of depression and associated anxiety
Catalog No. 50-310-8784 Supplier Selleck Chemical LLC Supplier No. S370125MG
Benactyzine hydrochloride is a centrally acting muscarinic antagonist It is used as an antidepressant in the treatment of depression and associated anxiety