Learn More
Medchemexpress LLC Benactyzine hydrochloride | 57-37-4 | 99.0% | 1 ML
SDP
Catalog No. 5000411169
Encompass_Preferred
Benactyzine hydrochloride is a competitive BChE inhibitor with a Ki of 0.01 mM. It is an anticholinergic drug and has potential for organophosphate poisoning research.
  • Competitive BChE inhibitor
  • Anticholinergic properties
  • Potential for organophosphate poisoning research
  • High purity of 99.0%
  • Available in various formulations
  • Optimized for stability with specific storage conditions
Catalog No. 50-004-11169 Supplier Medchemexpress LLC Supplier No. HYB1542A10MM/1ML
Benactyzine hydrochloride is a competitive BChE inhibitor with a Ki of 0.01 mM. It is an anticholinergic drug and has potential for organophosphate poisoning research.
  • Competitive BChE inhibitor
  • Anticholinergic properties
  • Potential for organophosphate poisoning research
  • High purity of 99.0%
  • Available in various formulations
  • Optimized for stability with specific storage conditions