Learn More
Medchemexpress LLC Benazepril | 86541-75-5 | 99.9% | C24H28N2O5 | 25 MG
SDP
Catalog No. 5000358000
Encompass_Preferred
Benazepril is an orally active angiotensin-converting enzyme (ACE) inhibitor that reduces angiotensin-II production. It inhibits oxidative stress and apoptosis via the PI3K/Akt signaling pathway. This compound improves diabetic nephropathy and decreases proteinuria, making it suitable for studies of hypertension, heart failure, and diabetic nephropathy.
  • Alleviates oxidative stress and inhibits cell apoptosis.
  • Improves diabetic nephropathy in Wistar male rats.
  • Decreases proteinuria, urea, creatinine, triglycerides, and total cholesterol levels.
  • Increases albumin level, diminishes kidney volume, and ameliorates pathological changes.
Catalog No. 50-003-58000 Supplier Medchemexpress LLC Supplier No. 25MGHYB0093
Benazepril is an orally active angiotensin-converting enzyme (ACE) inhibitor that reduces angiotensin-II production. It inhibits oxidative stress and apoptosis via the PI3K/Akt signaling pathway. This compound improves diabetic nephropathy and decreases proteinuria, making it suitable for studies of hypertension, heart failure, and diabetic nephropathy.
  • Alleviates oxidative stress and inhibits cell apoptosis.
  • Improves diabetic nephropathy in Wistar male rats.
  • Decreases proteinuria, urea, creatinine, triglycerides, and total cholesterol levels.
  • Increases albumin level, diminishes kidney volume, and ameliorates pathological changes.