Learn More
Cayman Chemical BenazeprIlat 10mg
SDP
Catalog No. 502985763
Encompass_Preferred
An ACE inhibitor; active metabolite of the prodrug benazepril
Catalog No. 50-298-5763 Supplier Cayman Chemical Supplier No. 2179610 MG
An ACE inhibitor; active metabolite of the prodrug benazepril