Learn More
Cayman Chemical Benazepril (hydrochloride), Purity: ≥98%, CAS: 86541-74-4, 50 mg
SDP
Catalog No. 502994778
Encompass_Preferred
A prodrug of benazeprilat; inhibits enzymatic activity of partially purified ACE isolated from rabbit lung (IC50 = 2 nM); decreases the triglyceride and total cholesterol levels in normotensive rats (30 mg/kg); decreases aortic atherosclerosis in cholesterol-fed rabbits (3 mg/kg per day); reduces blood pressure in spontaneously hypertensive rats (0.1-10 mg/kg per day); decreases proteinuria in cats with chronic kidney disease (0.5-1 mg/kg per day)
Catalog No. 50-299-4778 Supplier Cayman Chemical Supplier No. 2380450 MG
A prodrug of benazeprilat; inhibits enzymatic activity of partially purified ACE isolated from rabbit lung (IC50 = 2 nM); decreases the triglyceride and total cholesterol levels in normotensive rats (30 mg/kg); decreases aortic atherosclerosis in cholesterol-fed rabbits (3 mg/kg per day); reduces blood pressure in spontaneously hypertensive rats (0.1-10 mg/kg per day); decreases proteinuria in cats with chronic kidney disease (0.5-1 mg/kg per day)